missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSP Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547825
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 15% cross-reactivity with recombinant human MSP is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
MSP (or MST1) contains four kringle domains and a serine protease domain, similar to that found in hepatic growth factor. Despite the presence of the serine protease domain, the encoded protein may not have any proteolytic activity. The receptor for this protein is RON tyrosine kinase, which upon activation stimulates ciliary motility of ciliated epithelial lung cells. This protein is secreted and cleaved to form an alpha chain and a beta chain bridged by disulfide bonds.
Specifications
| MSP | |
| Polyclonal | |
| Unconjugated | |
| MST1 | |
| D3F15S2; D3F15S2h; D8h3f15s2; D9H3F15S2; DNF15S2; DNF15S2h; E2F transcription factor 2; E2F2; Hepatocyte growth factor-like protein; Hepatocyte growth factor-like protein alpha chain; Hepatocyte growth factor-like protein beta chain; hepatocyte growth factor-like protein homolog; Hgfl; macrophage stimulating 1; macrophage stimulating 1 (hepatocyte growth factor-like); macrophage stimulatory protein; Macrophage-stimulating protein; MSP; Mst1; NF15S2; Unknown (protein for MGC:137619) | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 15235 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P26928 | |
| MST1 | |
| CHO-derived recombinant mouse MSP/MST1 Gly19-Gly716 (Cys677Ala). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction