missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSP/MST1 Rabbit anti-Human, Mouse, Rat, Clone: 5R7X2, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £405.00
Specifications
| Antigen | MSP/MST1 |
|---|---|
| Clone | 5R7X2 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319115
|
Novus Biologicals
NBP3-15250-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347345
|
Novus Biologicals
NBP3-15250-100UL |
100 μg |
£405.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSP/MST1 Monoclonal antibody specifically detects MSP/MST1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| MSP/MST1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, NF15S2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MSP/MST1 (P26927). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 5R7X2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4485 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title