missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26053-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody does not react with talin or ezrin. It weakly reacts with radixin. It detects most of the proteolytic fragments of moesin.
Moesin, a member of the talin-4.1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures. This antibody does not react with talin or ezrin. It weakly reacts with radixin. It detects most of the proteolytic fragments of moesin.Specifications
| MSN | |
| Polyclonal | |
| Unconjugated | |
| MSN | |
| moesin, MSN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4478 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P26038 | |
| MSN | |
| MSN Fusion Protein Ag23531 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction