missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSMB Recombinant Rabbit Monoclonal Antibody (4L8B5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535454
This item is not returnable.
View return policy
Description
Immunogen sequence: MNVLLGSVVI FATFVTLCNA SCYFIPNEGV PGDSTRKCMD LKGNKHPINS EWQTDNCETC TCYETEISCC TLVSTPVGYD KDNCQRIFKK EDCKYIVVEK.
MSMB protein is a member of the immunoglobulin binding factor family. It is synthesized by the epithelial cells of the prostate gland and secreted into the seminal plasma. This protein has inhibin-like activity. It may have a role as an autocrine paracrine factor in uterine, breast and other female reproductive tissues. The expression of the encoded protein is found to be decreased in prostate cancer. Two alternatively spliced transcript variants encoding different isoforms are described for this gene. The use of alternate polyadenylation sites has been found for this gene.
Specifications
| MSMB | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08540, P08118 | |
| MSMB | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MSMB (P08118). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 4L8B5 | |
| Unconjugated | |
| MSMB | |
| beta-inhibin; beta-microseminoprotein; beta-MSP; HPC 13; HPC13; IGBF; immunoglobulin binding factor; Immunoglobulin-binding factor; microseminoprotein beta; microseminoprotein, beta; microseminoprotein, beta-; Msmb; MSP; MSPB; PIP; PN 44; PN44; Prostate secreted seminal plasma protein; prostate secretory protein of 94 amino acids; prostate secretory protein PSP94; prostatic inhibin protein; PRPS; PRSP; PSP; PSP 57; PSP57; PSP94; PSP-94; Seminal plasma beta-inhibin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17695, 4477 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction