missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSL2L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99028-50ul
This item is not returnable.
View return policy
Description
MSL2L1 Polyclonal antibody specifically detects MSL2L1 in Human samples. It is validated for Immunoprecipitation
Specifications
| MSL2L1 | |
| Polyclonal | |
| Immunoprecipitation 1-4 uL/mg of lysate | |
| FLJ10546, KIAA1585FLJ54913, male-specific lethal 2 homolog, male-specific lethal 2 homolog (Drosophila), Male-specific lethal 2-like 1, male-specific lethal 2-like 1 (Drosophila), Male-specific lethal-2 homolog 1, MSL-2, MSL2L1, MSL2-like 1, PHNPCC4, PMS2, RING finger protein 184msl-2, RNF184 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MSL2L1. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55167 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction