missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554826
This item is not returnable.
View return policy
Description
Immunogen sequence: FGSTERKTPV KKLAPEFSKV KTKTPKHSPI KEEPCGSLSE TVCKRELRSQ ETPEKPRSSV DTPPRLSTPQ KGPSTHPKEK AFSSEIEDLP YLSTTEMY Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.
Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at 'Lys-16' (H4K16ac) which is implicated in the formation of higher-order chromatin structure (PubMed:16227571). Greatly enhances MSL2 E3 ubiquitin ligase activity, promoting monoubiquitination of histone H2B at 'Lys-34' (H2BK34Ub) (PubMed:21726816). This modification in turn stimulates histone H3 methylation at 'Lys-4' (H3K4me) and 'Lys-79' (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1 (PubMed:21726816). In the MSL complex, acts as a scaffold to tether MSL3 and KAT8 together for enzymatic activity regulation (PubMed:22547026). [UniProt].
Specifications
| MSL1 | |
| Polyclonal | |
| Unconjugated | |
| MSL1 | |
| 2810017F12Rik; 4121402D02Rik; 4930463F05Rik; AA682082; hampin; hMSL1; male specific lethal 1 homolog; male-specific lethal 1 homolog; male-specific lethal 1 homolog (Drosophila); male-specific lethal 1-like 1; Male-specific lethal-1 homolog 1; Msl1; Msl-1; MSL1L1; MSL1-like 1; RGD1564778 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 339287 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68DK7 | |
| MSL1 | |
| Recombinant protein corresponding to Human MSL1. Recombinant protein control fragment (Product #RP-93537). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction