missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MSL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15662985
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15662985 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15662985 Supplier Invitrogen™ Supplier No. PA548690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with mouse based on sequence homology.

Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at 'Lys-16' (H4K16ac) which is implicated in the formation of higher-order chromatin structure (PubMed:16227571). Greatly enhances MSL2 E3 ubiquitin ligase activity, promoting monoubiquitination of histone H2B at 'Lys-34' (H2BK34Ub) (PubMed:21726816). This modification in turn stimulates histone H3 methylation at 'Lys-4' (H3K4me) and 'Lys-79' (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1 (PubMed:21726816). In the MSL complex, acts as a scaffold to tether MSL3 and KAT8 together for enzymatic activity regulation (PubMed:22547026). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MSL1
Gene Accession No. Q68DK7
Gene Alias 2810017F12Rik; 4121402D02Rik; 4930463F05Rik; AA682082; hampin; hMSL1; male specific lethal 1 homolog; male-specific lethal 1 homolog; male-specific lethal 1 homolog (Drosophila); male-specific lethal 1-like 1; Male-specific lethal-1 homolog 1; Msl1; Msl-1; MSL1L1; MSL1-like 1; RGD1564778
Gene Symbols MSL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 339-367 amino acids from the Central region of human MSL1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339287
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.