missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSK2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | MSK2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:20-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647071
|
Novus Biologicals
NBP2-93168-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643471
|
Novus Biologicals
NBP2-93168-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSK2 Polyclonal antibody specifically detects MSK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MSK2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 8986 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:20-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ribosomal protein S6 kinase, 90kDa, polypeptide 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 673-772 of human MSK2 (NP_003933.1). WLQDGSARSSPPLRTPDVLESSGPAVRSGLNATFMAFNRGKREGFFLKSVENAPLAKRRKQKLRSATASRRGSPAPANPGRAPVASKGAPRRANGPLPPS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title