missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSK1/RPS6KA5 Mouse anti-Human, Clone: PCRP-RPS6KA5-1A8, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | MSK1/RPS6KA5 |
|---|---|
| Clone | PCRP-RPS6KA5-1A8 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
Description
MSK1/RPS6KA5 Monoclonal specifically detects MSK1/RPS6KA5 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| MSK1/RPS6KA5 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 9252 | |
| IgG2b | |
| Protein A or G purified |
| PCRP-RPS6KA5-1A8 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RSKL, RSK-like protein kinase, S6K-alpha-5 | |
| Recombinant full-length human MSK1/RPS6KA5 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title