missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSK1/RPS6KA5 Antibody (PCRP-RPS6KA5-1A8), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08282JF549
This item is not returnable.
View return policy
Description
MSK1/RPS6KA5 Monoclonal antibody specifically detects MSK1/RPS6KA5 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| MSK1/RPS6KA5 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RSKL, RSK-like protein kinase, S6K-alpha-5 | |
| Recombinant full-length human MSK1/RPS6KA5 protein (Uniprot: O75582) | |
| 100 μg | |
| Protein Kinase, Signal Transduction | |
| 9252 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-RPS6KA5-1A8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction