missing translation for 'onlineSavingsMsg'
Learn More

MSK1/MSK2 [p Ser376, p Ser360] Antibody (1013F) [DyLight 488], Novus Biologicals™

Product Code. 30732663 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732663 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732663 Supplier Novus Biologicals Supplier No. FAB25181K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MSK1/MSK2 Monoclonal antibody specifically detects MSK1/MSK2 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSK1/MSK2
Applications Immunocytochemistry
Classification Monoclonal
Clone 1013F
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias 1110069D02Rik, 3110005L17Rik, 6330404E13Rik
Host Species Rabbit
Immunogen Phosphopeptide containing the human MSK1(S376) site., Accession # O75582
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.