Learn More
Eurofins DiscoverX MSK1 (2-end) Protein, Active
Description
MSK1 (mitogen- and stress-activated protein kinases-1, ribosomal protein S6 kinase, 90kDa, polypeptide 5) and MSK2 (mitogen- and stress-activated protein kinases-2, ribosomal protein S6 kinase, 90kDa, polypeptide 4) are activated in vitro by MAPK2/ERK2 or SAPK2/p38. Endogenous MSK is activated by UV radiation, oxidative stress, and other cell-damaging stimuli. MSK may mediate the growth factor and stress-induced activation of CREB.
GenBank AF074393
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 50% glycerol, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1 % 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 94.4 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥72% |
| Protein | MSK1 (2-end), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.