missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MSIA™ Microcolumns for the Freedom EVO Platform

Product Code. 15550290
Click to view available options
Description:
MSIA™ Streptavidin-EVO
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 15550290

Brand: Thermo Scientific™ 992STR96

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Next generation large molecule bio-analysis workflow for Freedom EVO Platform .

Next generation large molecule bio-analysis made easier by combining the Thermo Scientific™ innovative MSIA™ technology with Tecan advanced, robust and easy-to-use liquid handling. MSIA affinity microcolumns, designed for Freedom EVO platform (MCA 96 head), provide innovative enablement in the areas ranging from bio-analysis of therapeutic mAbs, antibody-drug conjugates (ADC), biotransformation [in-vivo modifications, drug-antibody ratio (DAR)], preclinical and clinical drug trials (PKPD, DMPK, and ADME) to biomarker and proteins analysis. This unique solution is designed to deliver the accuracy, reliability and automation you need to be successful.

Enabling workflows and technologies for biologics and biomarker research:

  • Robust Automation: Simply program the Freedom EVO to elute the analyte from complex biological samples and let the automated platform carry the load
  • Highly sensitive: Analyze proteins of interest down to femtomole level
  • Enhanced characteristics: Detects multiple protein variants for qualitative and quantitative bio-analysis in a single assay
  • Contaminant-free: proprietary molecular trapping microcolumn technology prevents matrix injection that would impact LC-MS function
  • High signal-to-noise ratio: Innovative sample interface technology reduces matrix loss, reagent carryover and non-specific binding
  • Time Savings: Microfluidic channels drive kinetic interactions significantly decreasing incubation time required to process the samples, while the MCA 96 head enables parallel processing of 96 samples
  • Reproducible: Consistent performance for assay reproducibility
  • Preserving samples: Small sample volumes (as low as 10 μL) and a reduced requirement for repeating experiments
  • MSIA Streptavidin EVO microcolumns provide end user flexibility to analyze their specific targets of interest
TRUSTED_SUSTAINABILITY

Specifications

Description MSIA™ Streptavidin-EVO
Volume (Metric) 10 μL
Storage Requirements 4°C
Format Rack
Product Line MSIA

NOTE: For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.