missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSI2 Recombinant Rabbit Monoclonal Antibody (24GB225)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557490
This item is not returnable.
View return policy
Description
MSI2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Msi2 (musashi homolog 2), also known as MSI2H, is a 328 amino acid protein that localizes to the cytoplasm and contains two RRM (RNA recognition motif) domains. Expressed ubiquitously at low levels, Msi2 functions as an RNA binding protein that, by regulating the expression of target mRNAs, is thought to play a role in the proliferation and maintenance of stem cells within the central nervous system. Msi2 is subject to posttranslational phosphorylation and is upregulated in response to brain injury, suggesting a role in healing and brain tissue regeneration. Chromosomal aberrations involving the Msi2 gene are associated with the progression of chronic myeloid leukemia. Multiple isoforms of Msi2 exist due to alternative splicing events.Tissue specificity: Ubiquitous; detected at low levels.
Specifications
| MSI2 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q920Q6, Q96DH6 | |
| MSI2 | |
| A synthesized peptide derived from human MSI2 (aa 200-280). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB225 | |
| Unconjugated | |
| MSI2 | |
| 1700105C15Rik; AI451722; AI563628; AW489193; MSI2; Msi2h; musashi homolog 2; musashi RNA binding protein 2; musashi RNA-binding protein 2; Musashi2; musashi-2; RGD1560397; RNA-binding protein Musashi homolog 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 124540, 360596, 76626 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction