missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSI-1436 lactate, MedChemExpress
Description
MSI-1436 lactate is a selective, non-competitive inhibitor of the enzyme protein-tyrosine phosphatase 1B (PTP1B), with an IC50 of 1 μM, 200-fold preference over TCPTP (IC50 of 224 μM).
Specifications
Specifications
| Chemical Name or Material | MSI-1436 lactate |
| CAS | 1309370-86-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C37H72N4O5S.7/2C3H6O3 |
| Quantity | 10 mg |
| Synonym | Trodusquemine lactate Aminosterol-1436 lactate |
| Solubility Information | DMSO : 54 mg/mL (53.99 mM; Need ultrasonic and warming) ∣0.1 M HCL : 50 mg/mL (49.99 mM; ultrasonic and adjust pH to 4 with HCl) |
| SMILES | NCCCNCCCCNCCCN[C@@H]1C[C@@](C[C@@H](O)[C@]2([H])[C@]3([H])CC[C@@]4(C)[C@@]2([H])CC[C@@]4([C@H](C)CC[C@@H](OS(=O)(O)=O)C(C)C)[H])([H])[C@]3(C)CC1.O=C(O)C(C)O.[3.5] |
| Molecular Weight (g/mol) | 1000.17 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?