missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSH6 Recombinant Rabbit Monoclonal Antibody (BLR117H)

Rabbit Recombinant Monoclonal Antibody
£128.00 - £385.00
Specifications
| Antigen | MSH6 |
|---|---|
| Clone | BLR117H |
| Concentration | 0.25 mg/mL |
| Content And Storage | 4°C |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17715799
![]() |
Invitrogen™
MA544425 |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17725799
|
Invitrogen™
MA544426 |
10 μL |
£128.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced malignant transformation and hence the prevention of skin cancer.Specifications
| MSH6 | |
| 0.25 mg/mL | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
| P52701 | |
| 2956 | |
| Peptide representing a region between residue 1310 and the C-terminus (residue 1360) of Human DNA mismatch repair protein Msh6. | |
| Antibody |
| BLR117H | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| MSH6 | |
| AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12 | |
| MSH6 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title