missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSH6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583010
This item is not returnable.
View return policy
Description
Immunogen sequence: MVENECEDPS QETITFLYKF IKGACPKSYG FNAARLANLP EEVIQKGHRK AREFEKMNQS LRLFREVCLA SERSTV Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 95%.
The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced malignant transformation and hence the prevention of skin cancer.
Specifications
| MSH6 | |
| Polyclonal | |
| Unconjugated | |
| MSH6 | |
| AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100360342, 17688, 2956 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P52701, P54276 | |
| MSH6 | |
| Recombinant protein corresponding to Human MSH6. Recombinant protein control fragment (Product #RP-95115). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction