missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSH6 Monoclonal Antibody (ZM006)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 394400
This item is not returnable.
View return policy
Description
MSH6 Monoclonal Antibody for Western Blot, ELISA
The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced malignant transformation and hence the prevention of skin cancer.
Specifications
| MSH6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P52701 | |
| MSH6 | |
| Recombinant protein derived from the N-terminal region of the human MSH6 protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 κ |
| ELISA, Western Blot | |
| ZM006 | |
| Unconjugated | |
| MSH6 | |
| AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2956 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction