missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH6 Antibody (rMSH6/8106), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £385.00
Specifications
| Antigen | MSH6 |
|---|---|
| Clone | rMSH6/8106 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605234
|
Novus Biologicals
NBP3-20350-100ug |
100 μg |
£385.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671475
|
Novus Biologicals
NBP3-20350-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MSH6 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, DNA Repair, Mismatch Repair | |
| 10mM PBS with 0.05% BSA | |
| 2956 | |
| IgG1 κ | |
| Protein A or G purified |
| rMSH6/8106 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein | |
| Recombinant fragment of human MSH6 protein (around aa 1-200) (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title