missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MSH6 Monoclonal antibody specifically detects MSH6 in Human, Mouse, Rat, Ferret, Monkey samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | MSH6 |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Regulatory Status | RUO |
| Research Discipline | Breast Cancer, Cancer, DNA Repair, Mismatch Repair |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?