missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94779-0.02ml
This item is not returnable.
View return policy
Description
MSH4 Polyclonal antibody specifically detects MSH4 in Human samples. It is validated for Western Blot
Specifications
| MSH4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| hMSH4, mutS (E. coli) homolog 4, mutS homolog 4 (E. coli), mutS protein homolog 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 697-936 of human MSH4 (NP_002431.2). QIMAQIGSYVPAEYSSFRIAKQIFTRISTDDDIETNSSTFMKEMKEIAYILHNANDKSLILIDELGRGTNTEEGIGICYAVCEYLLSLKAFTLFATHFLELCHIDALYPNVENMHFEVQHVKNTSRNKEAILYTYKLSKGLTEEKNYGLKAAEVSSLPPSIVLDAKEITTQITRQILQNQRSTPEMERQRAVYHLATRLVQTARNSQLDPDSLRIYLSNLKKKYKEDFPRTEQVPEKTEE | |
| 0.02 mL | |
| DNA Repair, Mismatch Repair | |
| 4438 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction