Learn More
MSH3 Rabbit, anti-Human, Clone: 1J23, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82845-3-RR-100UL
This item is not returnable.
View return policy
Description
The MSH3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MSH3. This antibody recognizes human antigen. The MSH3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| MSH3 | |
| 1J23 | |
| Divergent upstream protein, DUC1, DUG, DUP, hMSH3, Mismatch repair protein 1, MRP1, MSH3, mutS homolog 3 (E. coli) | |
| 100 μL | |
| 4437 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido