missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH3 Antibody (RM405), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89099
This item is not returnable.
View return policy
Description
MSH3 Monoclonal specifically detects MSH3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MSH3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 50% Glycerol, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM405 | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| Divergent upstream protein, DNA mismatch repair protein Msh3, DUC1, DUG, DUPMGC163306, hMSH3, Mismatch repair protein 1, MRP1MGC163308, mutS (E. coli) homolog 3, mutS homolog 3 (E. coli) | |
| A peptide corresponding to residues near the N-terminus of human MSH3 | |
| 100 μL | |
| DNA Repair, Mismatch Repair, Neuroscience | |
| 4437 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction