Learn More
MSH3 Antibody (CL14059) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24587AF350
Product Code. 30293521
  • £558.00 / 0.10mL
Description
MSH3 Monoclonal antibody specifically detects MSH3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSH3
Monoclonal
Alexa Fluor 350
Divergent upstream protein, DNA mismatch repair protein Msh3, DUC1, DUG, DUPMGC163306, hMSH3, Mismatch repair protein 1, MRP1MGC163308, mutS (E. coli) homolog 3, mutS homolog 3 (E. coli)
This antibody was developed using a synthetic peptide derived from P20585, with the exact immunogen sequence remaining proprietary.
0.1 mL
DNA Repair, Mismatch Repair, Neuroscience
4437
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL14059
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified