Learn More
Invitrogen™ MSH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579695
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human SH-SY5Y whole cell, human HEL whole cell, human 293T whole cell, human A549 whole cell, rat PC-12 whole cell, mouse Neuro-2a whole cell. IHC: Human Intestinal Cancer tissue. ICC/IF: U20S cell, SMMC cell. Flow: A431 cell.
Plays a role in neurogenesis and neuronal migration. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
Specifications
| MSH2 | |
| Polyclonal | |
| Unconjugated | |
| Msh2 | |
| AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17685, 4436, 81709 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P43246, P43247, P54275 | |
| Msh2 | |
| E.coli-derived human MSH2 recombinant protein (Position: Q337-N583). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.