missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH2 Mouse anti-Human, Clone: MSH2/2622, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07211-20UG
This item is not returnable.
View return policy
Description
MSH2 Monoclonal specifically detects MSH2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MSH2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin | |
| COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2 | |
| Recombinant fragment (around aa 327-427) of human MSH2 protein (exact sequence is proprietary) | |
| 20 μg | |
| Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors | |
| 4436 | |
| Store at 4°C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MSH2/2622 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction