missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56428
This item is not returnable.
View return policy
Description
MSH2 Polyclonal specifically detects MSH2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| MSH2 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| MSH2 | |
| A portion of amino acids 550-600 of human MSH2 was used as the immunogen. | |
| 0.1 mg | |
| Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors | |
| 4436 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, dog and S. cerevisiae, 94% in mouse and rat, and 76% in zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction