missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSDC 0160, MedChemExpress
Description
MSDC 0160 (Mitoglitazone) is a mitochondrial target of thiazolidinediones (mTOT)-modulating insulin sensitizer and a modulator of mitochondrial pyruvate carrier (MPC). MSDC 0160 is a thiazolidinedione (TZD) with antidiabetic and neuroprotective activities. MSDC 0160 has the potential for Alzheimer′s disease.
Specifications
Specifications
| Chemical Name or Material | MSDC 0160 |
| CAS | 146062-49-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H18N2O4S |
| Quantity | 50 mg |
| Synonym | Mitoglitazone CAY10415 |
| Solubility Information | DMSO : 33.33 mg/mL (89.98 mM; Need ultrasonic) |
| SMILES | O=C(N1)SC(CC2=CC=C(OCC(C3=NC=C(CC)C=C3)=O)C=C2)C1=O |
| Molecular Weight (g/mol) | 370.42 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?