missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSC Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA131456
This item is not returnable.
View return policy
Description
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX21330-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MyoD-E or musculin-E heterodimers bind avidly to consensus E box motifs, which are functionally important elements in the promoter regions of many musclespecific terminal differentiation genes. MyoD complexes potently induce transcriptional activation, while musculin complexes bind adjacent to MyoD DNA-binding regions to represses MyoD activity, which then results in the delayed expression of muscle-specific genes. Musculin is highly expressed in undifferentiated and proliferating myoblasts in culture, and its expression is down regulated during myogenesis and at the onset of terminal differentiation.
Specifications
| MSC | |
| Polyclonal | |
| Unconjugated | |
| MSC | |
| ABF1; ABF-1; Activated B-cell factor 1; activated B-cell factor 1, homolog of mouse musculin; activated B-cell factor-1; bHLHa22; Class A basic helix-loop-helix protein 22; Msc; Musculin; myogenic repressor; Myor | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 9242 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O60682 | |
| MSC | |
| Peptide with sequence C-PLKSDWNSLDGIIR, from the C Terminus. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction