Learn More
Thermo Scientific™ MSC-Advantage™ Class II Biological Safety Cabinets
Conserve energy with these cabinets, which combine smart design and extraordinary value with best-in-class energy efficiency, reliability and usability.
Brand: Thermo Scientific™ 51028226
8327.80 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
View more versions of this product
Description
Thermo Scientific™ MSC-Advantage™ Class II Biological Safety Cabinets (BSCs) combine safety and extraordinary value with energy efficiency, reliability and usability. With proprietary airflow design and energy-efficient DC motors, the MSC-Advantage BSCs use up to 68% less energy than cabinets with traditional AC motors. The cabinets provide both personal and product protection from biological hazards and contamination while affording users with a comfortable and ergonomic workspace. Cabinets are certified to EN 12469 safety standard.
Features:
- Available in 90cm, 120cm, 150cm and 180cm
- With microprocessor control
- Large front-panel display (easily visible from seated position) provides constant readout of downflow and inflow velocities and filter usage status
- Exclusive Performance Factor monitoring system advises when cabinet service is needed
- Proprietary window opens to 20cm working height, 53.5cm maximum height, easily lowers for interior cleaning
- Ergonomically angled window is sloped at 10° for better comfort and reduced operator fatigue
- Unique airflow technology and intelligent motor design dramatically improve energy efficiency, operation and serviceability; provide excellent sample protection
- SmartPort: T¨UV-approved, safe, clean method for vacuum tubing and cables routing method through the side wall of the biological safety cabinet
- Supply/exhaust air filter: HEPA H 14 EN 1822, 99.995% at 0.3μm particle size
Construction:
- Front-accessible components allow for quick change-outs of HEPA filters and other components Quiet operation at <55dBA enables a comfortable working environment
- Divided stainless-steel work tray removes easily for cleaning or autoclaving
- Standard armrests designed to prevent accidental blocking with coat sleeves
- Very large work surface (0.56m2 or 0.84m2) improves productivity
- Smooth components throughout the cabinet virtually eliminate risk of injury during routine cleaning, servicing or maintenance
Certifications/Compliance
- Fully compliant with the EN 12469 safety standard as independently tested and certified by TUV Nord
Ordering Information:
Accessories include programmable UV light, one-piece work tray, stand options, Ergolign saddle stool, free-standing floor shelf. Not available for sale in North America.
Specifications
EN 12469, by TüV Nord, LNE (0.9 and 1.2m). EN 12469, by TüV Nord (1.5 and 1.8m) | |
Biosafety Cabinet | |
2 Double, Left and Right Side | |
HEPA H 14 EN 1822, 99.995% at 0.3μm Particle | |
160 cm | |
TUV-approved | |
152 cm | |
230 V |
1.5 m | |
<59 dBA | |
62.9 in. | |
Energy efficient | |
<70 W | |
4.92 ft. | |
80 cm | |
280 W |