missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MSC 4381

Product Code. 30226367
Change view
Click to view available options
:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30226367 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30226367 Supplier Tocris Bioscience™ Supplier No. 8008/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective monocarboxylate transporter 4 (MCT4) inhibitor

MSC 4381 is a potent and selective monocarboxylate transporter 4 (MCT4) inhibitor (IC50 = 77 nM; Ki = 11 nM). Inhibits lactate efflux in MDA-MB-231 cells (IC50 = 1 nM) and reduces viability of cells expressing high levels of MCT4.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[2-[5-Chloro-2-[[(5-ethoxy-8-quinolinyl)sulfonyl]amino]phenyl]ethynyl]-4-methoxy-2-pyridinecarboxylic acid
CAS 2445185-57-7
Quantity 10 mg
Target Monocarboxylate Transporter Inhibitors
Molecular Formula C26H20ClN3O6S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.