missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ MsBM™ Mesangial Cell Growth Basal Medium

Product Code. 11655430
Click to view available options
Quantity:
500 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11655430

Brand: Lonza™ CC3147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a component in the full MsGM™ BulletKit™ (catalog number CC-3146) consisting of basal medium and supplements.

  • Basal medium.
  • Component in the full MsGM™ BulletKit™ (catalog number CC-3146), which consists of basal and supplements packaged together.
  • Specially formulated for the maintenance and expansion of renal mesangial cells.
  • Contains 5% serum.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mesangial Cells
Product Type Basal Medium
Quantity 500 mL
pH 7.5 to 8.0
Color Red
Content And Storage Store in dark at 2 to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.