missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSANTD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571431
This item is not returnable.
View return policy
Description
MSANTD4 Polyclonal Antibody for Western Blot
MSANTD4 is a protein coding gene.
Specifications
| MSANTD4 | |
| Polyclonal | |
| Unconjugated | |
| MSANTD4 | |
| 8430410K20Rik; DC25; KIAA1826; LOC511226 protein; Msantd4; Myb/SANT DNA binding domain containing 4 with coiled-coils; Myb/SANT-like DNA-binding domain containing 4 with coiled-coils; Myb/SANT-like DNA-binding domain-containing protein 4; Mysantd4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84437 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NCY6 | |
| MSANTD4 | |
| synthetic peptide directed towards the middle region of human MSANTD4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction