missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSANTD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571420
This item is not returnable.
View return policy
Description
MSANTD3 Polyclonal Antibody for Western Blot
MSANTD3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include identical protein binding.
Specifications
| MSANTD3 | |
| Polyclonal | |
| Unconjugated | |
| MSANTD3 | |
| 2410046L14Rik; 2810429P03Rik; 5730528L13Rik; C8H9orf30; C9orf30; L8; MSANTD3; Myb/SANT DNA binding domain containing 3; Myb/SANT-like DNA-binding domain containing 3; myb/SANT-like DNA-binding domain-containing protein 3; Mysantd3; RGD1308165; UPF0439 protein C9orf30; UPF0439 protein C9orf30 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 91283 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96H12 | |
| MSANTD3 | |
| synthetic peptide directed towards the C terminal region of human MSANTD3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction