missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSANTD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87846
This item is not returnable.
View return policy
Description
MSANTD3 Polyclonal specifically detects MSANTD3 in Human samples. It is validated for Western Blot.
Specifications
| MSANTD3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 91283 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| C9orf30, chromosome 9 open reading frame 30, FLJ34973, hypothetical protein LOC91283, L8, MGC17337, MSANTD3, Myb/SANT-like DNA-binding domain containing 3 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of human MSANTD3 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction