missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSANTD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144813
This item is not returnable.
View return policy
Description
Immunogen sequence: GYLVSPQAEK HRRARNWTDA EMRGLMLVWE EFFDELKQTK RNAKVYEKMA SKLFEMTGER RLGEEIKIKI TNMTFQ Antigen sequence identity: Mouse 99%; Rat 99%.
MSANTD1 is a protein coding gene.
Specifications
| MSANTD1 | |
| Polyclonal | |
| Unconjugated | |
| MSANTD1 | |
| A930005I04Rik; C3H4orf44; C4orf44; Gm144; MSANTD1; Myb/SANT DNA binding domain containing 1; Myb/SANT-like DNA-binding domain containing 1; myb/SANT-like DNA-binding domain-containing protein 1; RGD1565796; uncharacterized protein C4orf44 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 345222 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZTZ1 | |
| MSANTD1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 30-105. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction