Learn More
YMC Group YMC-Pack TMS, semi-preparative HPLC column 30 mm ID, 12 nm, S-5 µm, 100 x 30.0 mm
Brand: YMC Group TM12S05-1030WT
YMC-Pack TMS, semi-preparative HPLC column 30 mm ID, 12 nm, S-5 µm, 100 x 30.0 mm