missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | MS4A7 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
This gene encodes a member of the membrane-spanning 4A gene family, members of which are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns in hematopoietic cells and nonlymphoid tissues. This family member is associated with mature cellular function in the monocytic lineage, and it may be a component of a receptor complex involved in signal transduction. This gene is localized to 11q12, in a cluster of other family members. At least four alternatively spliced transcript variants encoding two distinct isoforms have been observed.Specifications
| MS4A7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9GZW8 | |
| 58475 | |
| MS4A7 Fusion Protein Ag2442 | |
| Primary | |
| -20°C | |
| MS4A7 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 4SPAN2, CD20 antigen like 4, CD20L4, CFFM4, MS4A7, MS4A8 | |
| MS4A7 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title