missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93062-0.02ml
This item is not returnable.
View return policy
Description
MS4A7 Polyclonal antibody specifically detects MS4A7 in Mouse samples. It is validated for Western Blot
Specifications
| MS4A7 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| CD20/FC-epsilon-RI-beta family member 4, CD20L4MGC22368, CFFM4CD20 antigen-like 4, Four-span transmembrane protein 2, high affinity immunoglobulin epsilon receptor beta subunit, membrane-spanning 4-domains subfamily A member 7, membrane-spanning 4-domains, subfamily A, member 7,4SPAN2, MS4A8 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human MS4A7 (NP_067024.1). SLSIISGKQSTKPFDLSSLTSNAVSSVTAGAGLFLLADSMVALRTASQHCGSEMDYLSSLPYSEYYYPIYEIKDCLLTSVSLTGVLVVMLIFTVLELLLAA | |
| 0.02 mL | |
| Cell Biology | |
| 58475 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction