missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MS4A7 Polyclonal antibody specifically detects MS4A7 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | MS4A7 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD20/FC-epsilon-RI-beta family member 4, CD20L4MGC22368, CFFM4CD20 antigen-like 4, Four-span transmembrane protein 2, high affinity immunoglobulin epsilon receptor beta subunit, membrane-spanning 4-domains subfamily A member 7, membrane-spanning 4-domains, subfamily A, member 7,4SPAN2, MS4A8 |
| Host Species | Rabbit |
| Immunogen | membrane-spanning 4-domains, subfamily A, member 7. (Uniprot# Q9GZW8) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?