missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MS4A6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572732
This item is not returnable.
View return policy
Description
MS4A6A Polyclonal Antibody for Western Blot, IHC (P), ELISA
May be involved in signal transduction as a component of a multimeric receptor complex.
Specifications
| MS4A6A | |
| Polyclonal | |
| Unconjugated | |
| MS4A6A | |
| 4SPAN3; 4SPAN3.2; CD20 antigen-like 3; CD20L3; CD20-like precusor; CDA01; four-span transmembrane protein 3; four-span transmembrane protein 3.1; four-span transmembrane protein 3.2; HAIRB-iso; membrane spanning 4-domains A6A; membrane-spanning 4-domains subfamily A member 6A; membrane-spanning 4-domains subfamily A member 6D; membrane-spanning 4-domains, subfamily A, member 11; membrane-spanning 4-domains, subfamily A, member 6A; membrane-spanning 4-domains, subfamily A, member 6A, isoform 2; Ms4a11; MS4A6; MS4A6A; MS4A6A-polymorph; MST090; MSTP090 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361735, 64231 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9H2W1 | |
| MS4A6A | |
| MS4A6A antibody for a 19 amino acid synthetic peptide near the amino terminus of human MS4A6A. The immunogen is within the first 50 amino acids of MS4A6A. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction