missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A6A, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00064231-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MTSQPVPNETIIVPPSNVINFSQAEKPEPTNQGQDSLKKHLHAEIKVIGTIQILCGMMVLSLGIILASASFSPNFTQVTSTLLNSAYPFIGPFFFIISGSLSIATEKRLTKLLVHSSLVGSILSALSALVGFIILSVKQATLNPASLQCELDKNNIPTRSYVSYFYHDSLYTTDCYTAKASLAGSLSLMLICTLLEFCLAVLTAVLRWKQAYSDFPGSVLFLPHSYIGNSGMSSKMTHDCGYEELLTSSpecifications
| MS4A6A | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC022854 | |
| MS4A6A | |
| MS4A6A (AAH22854, 1 a.a. ∼ 248 a.a) full-length human protein. | |
| RUO | |
| 64231 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| membrane-spanning 4-domains, subfamily A, member 6A | |
| 4SPAN3/4SPAN3.2/CD20L3/CDA01/MGC131944/MGC22650/MS4A6/MST090/MSTP090 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction