missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MS4A6A Polyclonal antibody specifically detects MS4A6A in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | MS4A6A |
| Applications | Immunohistochemistry |
| Purification Method | Peptide affinity purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?