missing translation for 'onlineSavingsMsg'
Learn More

MS4A4A Antibody (3F2), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18340293 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340293 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340293 Supplier Novus Biologicals Supplier No. NBP312045JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MS4A4A Monoclonal antibody specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A4A
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 3F2
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7
Host Species Rabbit
Immunogen This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51338
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.