missing translation for 'onlineSavingsMsg'
Learn More

MS4A3 Antibody, Novus Biologicals™

Product Code. 18338187 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338187 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338187 Supplier Novus Biologicals Supplier No. H00000932B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

MS4A3 Polyclonal antibody specifically detects MS4A3 in Human samples. It is validated for Western Blot, Flow Cytometry, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A3
Applications Western Blot, Flow Cytometry, Proximity Ligation Assay
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH08487
Gene Alias CD20 antigen homolog, CD20 antigen-like protein, CD20LIgE receptor beta subunit, hematopoietic cell 4 transmembrane protein, Hematopoietic-specific transmembrane protein 4, HTm4, HTM4IgE receptor beta chain, membrane-spanning 4-domains subfamily A member 3, membrane-spanning 4-domains, subfamily A, member 3 (hematopoieticcell-specific)
Host Species Mouse
Immunogen MS4A3 (AAH08487, 1 a.a. - 214 a.a.) full-length human protein. MASHEVDNAELGSASARGTPGSEAGPEELNTSVYQPIDGSPDYQKAKLQVLGAIQILNAAMILALGVFLGSLQYPYHFQKHFFFFTFYTGYPIWGAVFFCSSGTLSVVAGIKPTRTWIQNSFGMNIASATIALVGTAFLSLNIAVNIQSLRSCHSSSESPDLCNYMGSISNGMVSLLLILTLLELCVTISTIAMWCNANCCNSREEISSPPNSV
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 932
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.