missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A3 Antibody (489433), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6247APCCY7
This item is not returnable.
View return policy
Description
MS4A3 Monoclonal antibody specifically detects MS4A3 in Human samples. It is validated for Flow Cytometry
Specifications
| MS4A3 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 932 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 489433 | |
| Flow Cytometry | |
| CD20 antigen homolog, CD20 antigen-like protein, CD20LIgE receptor beta subunit, hematopoietic cell 4 transmembrane protein, Hematopoietic-specific transmembrane protein 4, HTm4, HTM4IgE receptor beta chain, membrane-spanning 4-domains subfamily A member 3, membrane-spanning 4-domains, subfamily A, member 3 (hematopoieticcell-specific) | |
| NS0 mouse myeloma cell line transfected with human MS4A3, Accession # NP_006129 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction