missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A15 Antibody (3D2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00219995-M05
This item is not returnable.
View return policy
Description
MS4A15 Monoclonal antibody specifically detects MS4A15 in Human samples. It is validated for ELISA
Specifications
| MS4A15 | |
| Monoclonal | |
| Unconjugated | |
| AAH31610 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 219995 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| ELISA | |
| 3D2 | |
| In 1x PBS, pH 7.4 | |
| FLJ34527, membrane-spanning 4-domains subfamily A member 15, membrane-spanning 4-domains, subfamily A, member 15, MGC35295 | |
| MGC35295 (AAH31610.1, 1 a.a. ∽ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVRRGHVGIFFIEGGVPFWGGACFIISGSLSVAAEKNHTSCLVRSSLGTNILSVMAAFAGTAILLMDFGVTNRDVDRGYLAVLTIFTVLEFFTAVIAMHFGCQAIHAQASAPVIFLPNAFSADFNIPSPAASAPPAYDNVAYAQGVV | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction