Learn More
Invitrogen™ MS4A14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139841
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DQLQFVLQEE FSSDDSTTNA QSVIFGGYAF FKLTLSRSPL VSQPGNKGRE FVPDEQKQSI LPSPKFSEEE IEPLPP Highest antigen sequence indentity to the following orthologs: Mouse - 40%, Rat - 37%.
May be involved in signal transduction as a component of a multimeric receptor complex.
Specifications
| MS4A14 | |
| Polyclonal | |
| Unconjugated | |
| MS4A14 | |
| membrane spanning 4-domains A14; membrane-spanning 4-domains subfamily A member 14; membrane-spanning 4-domains, subfamily A, member 14; membrane-spanning 4-domains, subfamily A, member 16; MS4A13; MS4A13 protein; MS4A14; MS4A16; NYD-SP21; testes development-related NYD-SP21; testis development protein NYD-SP21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JA4 | |
| MS4A14 | |
| Recombinant protein corresponding to Human MS4A14. Recombinant protein control fragment (Product #RP-108964). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.