missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A14 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33254-100ul
This item is not returnable.
View return policy
Description
MS4A14 Monoclonal antibody specifically detects MS4A14 in Rat samples. It is validated for ELISA,Western Blot
Specifications
| MS4A14 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| DKFZp434H092, FLJ32856, membrane-spanning 4-domains subfamily A member 14, membrane-spanning 4-domains, subfamily A, member 14, membrane-spanning 4-domains, subfamily A, member 16, MGC104289, MGC49828, MS4A13, MS4A13 protein, MS4A16, NYD-SP21, testes development-related NYD-SP21, Testis development protein NYD-SP21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 150-280 of human MS4A14 (Q96JA4).,, Sequence:, FSRTLFIVLFFLPSDVTQNSEQPAPEENDQLQFVLQEEFSSDDSTTNAQSVIFGGYAFFKLTLSRSPLVSQPGNKGREFVPDEQKQSILPSPKFSEEEIEPLPPTLEKKPSENMSIQLDSTFKQMKDEDLQ | |
| 100 μL | |
| Primary | |
| Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84689 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction