missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13293-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cell surface protein found primarily in the apical membrane of colonocytes. Silencing of this gene in colon cancer cells inhibits the proliferation, cell motility, and chemotactic invasion of cells. This gene is part of a cluster of similar genes found on chromosome 11. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| MS4A12 | |
| Polyclonal | |
| Unconjugated | |
| MS4A12 | |
| Ms4a10, MS4A12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 381213, 54860 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NXJ0 | |
| Ms4a12 | |
| MS4A12 Fusion Protein Ag3968 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction